missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide, 98%

Catalog No. AAA1888230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1888230 250 g
AAA1888209 10 g
AAA1888218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1888230 Supplier Thermo Scientific Chemicals Supplier No. A1888230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52509-14-5 | C22H22BrO2P | 429.29 g/mol

It finds its applications as a reagent for Wittig olefinations with introduction of a 1,3-dioxolane moiety. It is applied as a reactant for the preparation of a ratiometric fluorescent probe for specific detection of cysteine over homocysteine and glutathione, for microwave-assisted synthesis of KN-93 as inhibitor of calmodulin kinase II, for the preparation of fluorinated spirobenzofuran piperidines as s1 receptor ligands, in the synthesis and antitumor agents, in regio-selective preparation of indole derivatives via rhodium-catalyzed domino hydroformylation/indolization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its applications as a reagent for Wittig olefinations with introduction of a 1,3-dioxolane moiety. It is applied as a reactant for the preparation of a ratiometric fluorescent probe for specific detection of cysteine over homocysteine and glutathione, for microwave-assisted synthesis of KN-93 as inhibitor of calmodulin kinase II, for the preparation of fluorinated spirobenzofuran piperidines as s1 receptor ligands, in the synthesis and antitumor agents, in regio-selective preparation of indole derivatives via rhodium-catalyzed domino hydroformylation/indolization.

Solubility
Soluble in water (20°C).

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 52509-14-5
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00011966
InChI Key FRHRVQQUICVJDG-UHFFFAOYSA-M
Synonym 1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy
PubChem CID 2724194
IUPAC Name 1,3-dioxolan-2-ylmethyl(triphenyl)phosphanium;bromide
SMILES [Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 186°C to 189°C
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (20°C).
Formula Weight 429.3
Percent Purity 98%
Chemical Name or Material (1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 257-977-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only