accessibility menu, dialog, popup

UserName

H3CCH3NH2

CAS RN 52430-81-6

IUPAC Name: 4-(propan-2-yl)cyclohexan-1-amine
Synonyms: 4-isopropylcyclohexan-1-amine
Molecular Weight (g/mol): 141.26
Molecular Formula: C9H19N
InChi Key: MFRKYEJMLQUSJX-UHFFFAOYSA-N
SMILES: CC(C)C1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results