accessibility menu, dialog, popup

UserName

CAS RN 52415-29-9

IUPAC Name: 6-bromo-1H-indole
Synonyms: 6-bromo-1H-indole
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: MAWGHOPSCKCTPA-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CNC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)