accessibility menu, dialog, popup

UserName

CAS RN 52196-74-4

CH3OOCH3Cl

CAS RN 52196-74-4

IUPAC Name: 2-chloro-1,4-diethoxybenzene
Synonyms: 2-chloro-1,4-diethoxybenzene
Molecular Weight (g/mol): 200.66
Molecular Formula: C10H13ClO2
InChi Key: ZIMKMIAIVORSSX-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(OCC)C(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results