accessibility menu, dialog, popup

UserName

CAS RN 521-24-4

IUPAC Name: sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate
Synonyms: folin's reagent
Molecular Weight (g/mol): 260.19
Molecular Formula: C10H5NaO5S
InChi Key: UBLXEEBHYISRFM-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC(=O)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)