accessibility menu, dialog, popup

UserName

FFFSOOOFFFSOOOFFFSOOOY

CAS RN 52093-30-8

IUPAC Name: yttrium(3+) tritrifluoromethanesulfonate
Synonyms: yttrium(3+) tritriflate
Molecular Weight (g/mol): 536.1
Molecular Formula: C3F9O9S3Y
InChi Key: JPJIEXKLJOWQQK-UHFFFAOYSA-K
SMILES: [Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results