accessibility menu, dialog, popup

UserName

CAS RN 520-26-3

IUPAC Name: (2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: hesperidin (S)-(--)-hesperidin cirantin hesperidoside 橙皮苷
Molecular Weight (g/mol): 610.57
Molecular Formula: C28H34O15
InChi Key: QUQPHWDTPGMPEX-QJBIFVCTSA-N
SMILES: COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)