accessibility menu, dialog, popup

UserName

CAS RN 520-18-3

IUPAC Name: 3,5,7-trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Synonyms: kaempherol kempferol campherol trifolitin pelargidenolon 1497 rhamnolutein ccris 41 pelargidenolon robigenin indigo yellow
Molecular Weight (g/mol): 286.24
Molecular Formula: C15H10O6
InChi Key: IYRMWMYZSQPJKC-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)