missing translation for 'onlineSavingsMsg'
Learn More

Kaempferol, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014425
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014425 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014425 Supplier Merck Emd Millipore Supplier No. 9635310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kaempferol is a bioflavonoid and a main component of the extract of Ginkgo biloba. It is also present in a variety of fruits and vegetables. It may serve as a potent candidate in the treatment of postmenopausal bone loss.

Featured Industries: Food and Beverages

Specifications

CAS 520-18-3
Physical Form Neat
Linear Formula C15H10O6
MDL Number MFCD00016938
Synonym 3,4′,5,7-Tetrahydroxyflavone; 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4 H-1-benzopyran-4-one; Robigenin
Type Analytical Standard
Quantity 10 mg
Formula Weight 286.24
Percent Purity ≥99% (HPLC)
Grade Analytical Standard