missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kaempferol, 97%

Catalog No. AC434731000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434731000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434731000 Supplier Thermo Scientific Chemicals Supplier No. 434731000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 520-18-3 | C15H10O6 | 286.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 520-18-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.24
MDL Number MFCD00016938
InChI Key IYRMWMYZSQPJKC-UHFFFAOYSA-N
Synonym kaempferol, kaempherol, kempferol, populnetin, rhamnolutein, robigenin, trifolitin, pelargidenolon, rhamnolutin, swartziol
PubChem CID 5280863
ChEBI CHEBI:28499
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 282°C to 286°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15,5322
Formula Weight 286.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Kaempferol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-287-6

RUO – Research Use Only