CAS RN 51795-97-2
IUPAC Name:
[(6-bromohexyl)oxy]benzene
Synonyms:
[(6-bromohexyl)oxy]benzene
Molecular Weight (g/mol):
257.17
Molecular Formula:
C12H17BrO
InChi Key:
QOLSAZPSEJXYSH-UHFFFAOYSA-N
SMILES:
BrCCCCCCOC1=CC=CC=C1
1
–
1
of
1
results
1
6-Phenoxyhexyl Bromide 96.0+%, TCI America™
CAS: 51795-97-2 Molecular Formula: C12H17BrO Molecular Weight (g/mol): 257.171 MDL Number: MFCD00039205 InChI Key: QOLSAZPSEJXYSH-UHFFFAOYSA-N Synonym: 6-Bromohexyl Phenyl Ether PubChem CID: 142878 IUPAC Name: 6-bromohexoxybenzene SMILES: C1=CC=C(C=C1)OCCCCCCBr