missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxyhexyl Bromide 96.0+%, TCI America™

Catalog No. P18485G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18485G Supplier TCI America Supplier No. P18485G
Only null left

Chemical Identifiers

CAS 51795-97-2
Molecular Formula C12H17BrO
Molecular Weight (g/mol) 257.171
MDL Number MFCD00039205
InChI Key QOLSAZPSEJXYSH-UHFFFAOYSA-N
Synonym 6-Bromohexyl Phenyl Ether
PubChem CID 142878
IUPAC Name 6-bromohexoxybenzene
SMILES C1=CC=C(C=C1)OCCCCCCBr

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 257.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Phenoxyhexyl Bromide