accessibility menu, dialog, popup

UserName

CAS RN 51729-83-0

IUPAC Name: methyl propan-2-yl carbonate
Synonyms: isopropyl methyl carbonate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: RCIJMMSZBQEWKW-UHFFFAOYSA-N
SMILES: COC(=O)OC(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results