CAS RN 51719-65-4
IUPAC Name:
2-(3,5-dichlorophenyl)acetic acid
Synonyms:
(3,5-dichlorophenyl)acetic acid
Molecular Weight (g/mol):
205.03
Molecular Formula:
C8H6Cl2O2
InChi Key:
RERINLRFXYGZEE-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(Cl)=CC(Cl)=C1
1
–
1
of
1
results
1
Promotions Available
3,5-Dichlorophenylacetic acid, 95%
3,5-Dichlorophenylacetic acid, 95%
CAS: 51719-65-4 Molecular Formula: C8H6Cl2O2 Molecular Weight (g/mol): 205.034 MDL Number: MFCD01764671 InChI Key: RERINLRFXYGZEE-UHFFFAOYSA-N Synonym: 2-3,5-dichlorophenyl acetic acid,3,5-dichlorophenylacetic acid,benzeneacetic acid, 3,5-dichloro,3,5-dichlorobenzeneacetic acid,3,5-dichlorophenyl acetic acid,3,5-dichloro-phenyl-acetic acid,2-3,5-dichlorophenyl-acetic acid,3,5-dichlorophenylaceticacid,pubchem24116,3,5-dichlorphenylacetic acid PubChem CID: 6452487 IUPAC Name: 2-(3,5-dichlorophenyl)acetic acid SMILES: C1=C(C=C(C=C1Cl)Cl)CC(=O)O