CAS RN 517-51-1
Rubrene, 99%
CAS: 517-51-1 Molecular Formula: C42H28 Molecular Weight (g/mol): 532.67 MDL Number: MFCD00003703 InChI Key: YYMBJDOZVAITBP-UHFFFAOYSA-N Synonym: rubrene,5,6,11,12-tetraphenylnaphthacene,rubren,naphthacene, 5,6,11,12-tetraphenyl,rubrene, powder,acmc-209kvj,5,11,12-tetraphenyltetracene,5,11,12-tetraphenylnaphthacene,naphthacene,6,11,12-tetraphenyl PubChem CID: 68203 IUPAC Name: 5,6,11,12-tetraphenyltetracene SMILES: C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
Rubrene, 97%
CAS: 517-51-1 Molecular Formula: C42H28 Molecular Weight (g/mol): 532.686 MDL Number: MFCD00003703 InChI Key: YYMBJDOZVAITBP-UHFFFAOYSA-N Synonym: rubrene,5,6,11,12-tetraphenylnaphthacene,rubren,naphthacene, 5,6,11,12-tetraphenyl,rubrene, powder,acmc-209kvj,5,11,12-tetraphenyltetracene,5,11,12-tetraphenylnaphthacene,naphthacene,6,11,12-tetraphenyl PubChem CID: 68203 IUPAC Name: 5,6,11,12-tetraphenyltetracene SMILES: C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
5,6,11,12-Tetraphenylnaphthacene 98.0+%, TCI America™
CAS: 517-51-1 Molecular Formula: C42H28 Molecular Weight (g/mol): 532.686 MDL Number: MFCD00003703 InChI Key: YYMBJDOZVAITBP-UHFFFAOYSA-N Synonym: rubrene,5,6,11,12-tetraphenylnaphthacene,rubren,naphthacene, 5,6,11,12-tetraphenyl,rubrene, powder,acmc-209kvj,5,11,12-tetraphenyltetracene,5,11,12-tetraphenylnaphthacene,naphthacene,6,11,12-tetraphenyl PubChem CID: 68203 IUPAC Name: 5,6,11,12-tetraphenyltetracene SMILES: C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
5,6,11,12-Tetraphenylnaphthacene (purified by sublimation) 99.0+%, TCI America™
CAS: 517-51-1 Molecular Formula: C42H28 Molecular Weight (g/mol): 532.686 MDL Number: MFCD00003703 InChI Key: YYMBJDOZVAITBP-UHFFFAOYSA-N Synonym: rubrene,5,6,11,12-tetraphenylnaphthacene,rubren,naphthacene, 5,6,11,12-tetraphenyl,rubrene, powder,acmc-209kvj,5,11,12-tetraphenyltetracene,5,11,12-tetraphenylnaphthacene,naphthacene,6,11,12-tetraphenyl PubChem CID: 68203 IUPAC Name: 5,6,11,12-tetraphenyltetracene SMILES: C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8