accessibility menu, dialog, popup

UserName

(S)(R)OOHOHHOOHHOH

CAS RN 517-28-2

IUPAC Name: (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Synonyms: (+)-hematoxylin
Molecular Weight (g/mol): 302.28
Molecular Formula: C16H14O6
InChi Key: WZUVPPKBWHMQCE-WKTCHCBJNA-N
SMILES: [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
Quantity 
  • (4)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
  • (2)
Color 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)