missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin, MP Biomedicals™

Catalog No. ICN10192225
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10192225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10192225 Supplier MP Biomedicals Inc Supplier No. 0210192225

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym hematoxylin, hydroxybrasilin
PubChem CID 45029742
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Color Red-Tan
Quantity 25 g
Formula Weight 302.3
Physical Form Powder
Chemical Name or Material Hematoxylin