missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin (Certified Biological Stain), Fisher Chemical™

Catalog No. H34525
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H34525 25 g Glass Bottle
H345100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H34525 Supplier Thermo Fisher Scientific Supplier No. H345 25
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in histology and cytology

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black 1, Hydroxybrasilin
PubChem CID 45029742
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Melting Point 100°C
Bio. Stain Comm. Certification Pass Test
Packaging Glass Bottle
Color Index No. 75290
Color Amber
Quantity 25 g
Physical Form Powder
Formula Weight 302.28
Grade Certified Biological Stain
Chemical Name or Material Hematoxylin