CAS RN 5160-02-1
CAS RN 5160-02-1
IUPAC Name:
barium(2+) bis(5-chloro-4-methyl-2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}benzene-1-sulfonate)
Synonyms:
C.I. pigment red 53
Molecular Weight (g/mol):
888.93
Molecular Formula:
C34H24BaCl2N4O8S2
InChi Key:
JNIGYQOBELCEIZ-MAKDUZDQSA-L
SMILES:
[Ba++].CC1=CC(N\N=C2/C(=O)C=CC3=CC=CC=C23)=C(C=C1Cl)S([O-])(=O)=O.CC1=CC(N\N=C2/C(=O)C=CC3=CC=CC=C23)=C(C=C1Cl)S([O-])(=O)=O
1
–
1
of
1
results
1
Lake Red CBA, TCI America™
CAS: 5160-02-1 Molecular Formula: C34H24BaCl2N4O8S2 Molecular Weight (g/mol): 888.93 MDL Number: MFCD01941571 InChI Key: JNIGYQOBELCEIZ-MAKDUZDQSA-L Synonym: 1-(4-Chloro-o-sulfo-5-tolylazo)-2-naphthol Barium Salt PubChem CID: 131842502 IUPAC Name: barium;5-chloro-4-methyl-2-[(2Z)-2-(2-oxonaphthalen-1-ylidene)hydrazinyl]benzenesulfonic acid SMILES: CC1=CC(=C(C=C1Cl)S(=O)(=O)O)NN=C2C(=O)C=CC3=CC=CC=C32.CC1=CC(=C(C=C1Cl)S(=O)(=O)O)NN=C2C(=O)C=CC3=CC=CC=C32.[Ba]