accessibility menu, dialog, popup

UserName

CAS RN 5159-41-1

IUPAC Name: (2-iodophenyl)methanol
Synonyms: 2-jod-benzylalkohol
Molecular Weight (g/mol): 234.04
Molecular Formula: C7H7IO
InChi Key: WZCXOBMFBKSSFA-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)