accessibility menu, dialog, popup

UserName

CAS RN 5153-40-2

IUPAC Name: 1-bromo-2,3,4,5,6-pentamethylbenzene
Synonyms: 1-bromo-2,3,4,5,6-pentamethylbenzene
Molecular Weight (g/mol): 227.15
Molecular Formula: C11H15Br
InChi Key: XPDQRULPGCFCLX-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(Br)C(C)=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)