missing translation for 'onlineSavingsMsg'
Learn More

Bromopentamethylbenzene 97.0+%, TCI America™

Catalog No. B41545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41545G 5 g
B415425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41545G Supplier TCI America Supplier No. B41545G

Chemical Identifiers

CAS 5153-40-2
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.15
MDL Number MFCD00013523
InChI Key XPDQRULPGCFCLX-UHFFFAOYSA-N
Synonym bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene
PubChem CID 78831
IUPAC Name 1-bromo-2,3,4,5,6-pentamethylbenzene
SMILES CC1=C(C)C(C)=C(Br)C(C)=C1C

Specifications

Melting Point 162°C
Color White-Yellow
Boiling Point 290°C
Quantity 5 g
Formula Weight 227.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bromopentamethylbenzene