accessibility menu, dialog, popup

UserName

CH3BrF

CAS RN 51436-99-8

IUPAC Name: 4-bromo-2-fluoro-1-methylbenzene
Synonyms: toluene,2-fluoro-4-bromo
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: YZFVUQSAJMLFOZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1F
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)