missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorotoluene, 99%

Catalog No. AAA1589122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1589122 100 g
AAA1589106 5 g
AAA1589114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1589122 Supplier Thermo Scientific Chemicals Supplier No. A1589122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51436-99-8 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51436-99-8
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00013551
InChI Key YZFVUQSAJMLFOZ-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorotoluene, 2-fluoro-4-bromotoluene, benzene, 4-bromo-2-fluoro-1-methyl, toluene,2-fluoro-4-bromo, 4-bromo-2-fluoro-1-methyl-benzene, pubchem1601, acmc-1an2v, 2-fluoro-4-bromo toluene, 4-bromo-2-fluoro-toluene
PubChem CID 171040
IUPAC Name 4-bromo-2-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)F

Specifications

Density 1.492
Boiling Point 66°C to 68°C (8 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.529
Quantity 100 g
Beilstein 1859028
Formula Weight 189.03
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-fluorotoluene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 257-201-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only