accessibility menu, dialog, popup

UserName

CdOOO

CAS RN 513-78-0

IUPAC Name: cadmium(2+) carbonate
Synonyms: cadmium carbonate
Molecular Weight (g/mol): 172.42
Molecular Formula: CCdO3
InChi Key: GKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES: [Cd++].[O-]C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (10)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (9)
  • (4)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)