missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Carbonate, Pure

Catalog No. AC212322500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC212322500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC212322500 Supplier Thermo Scientific Chemicals Supplier No. 212322500

CAS: 513-78-0 | CCdO3 | 172.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
IUPAC Name cadmium(2+) carbonate
SMILES [Cd++].[O-]C([O-])=O

Specifications

Melting Point 343°C to 412°C
Color White
Physical Form Solid
Quantity 250 g
Assay Percent Range 60.0 to 63.0% (Cd)
Linear Formula CdCO3
UN Number 2570
Fieser 04,67
Merck Index 15,1621
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted acids and in concentrated, solutions of nh4-salts
Formula Weight 172.41
Grade Pure
Concentration or Composition (by Analyte or Components) >95%
Odor Odorless
Appearance Powder
Chemical Name or Material Cadmium Carbonate, Pure
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs thr
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISO
DOTInformation Hazard Class: 6.1
EINECSNumber 208-168-9
Recommended Storage Keep containers tightly closed in a dry, cool and well-ventilated place

RUO – Research Use Only