missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Carbonate, Powder, Reagent, 97%, Spectrum™ Chemical

Catalog No. 18601178
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601178 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18601178 Supplier Spectrum Chemical Mfg Cor Supplier No. C1020500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C1020, 513-78-0, CdCO3

Spectrum™ Chemical Cadmium Carbonate, Powder, Reagent , also known as Otavite, a naturally occurring mineral. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
IUPAC Name cadmium(2+) carbonate
SMILES [Cd++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
UN Number UN2570
Percent Purity 97%
Grade Reagent
Loss on Ignition 25.5-27.5%
Packaging Poly Bottle