accessibility menu, dialog, popup

UserName

OCNNCO

CAS RN 5124-30-1

IUPAC Name: 1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
Synonyms: HMDI hydrogenated mdi hylene W
Molecular Weight (g/mol): 262.35
Molecular Formula: C15H22N2O2
InChi Key: KORSJDCBLAPZEQ-UHFFFAOYSA-N
SMILES: O=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)