missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyrazine-2-carboxylic Acid 4-Oxide 98.0+%, TCI America™

Catalog No. M20535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20535G 5 g
M20531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20535G Supplier TCI America Supplier No. M20535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51037-30-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00865757
InChI Key DNRXJHATQULEHC-UHFFFAOYSA-N
Synonym acipimox, olbetam, olbemox, 5-carboxy-2-methylpyrazine 1-oxide, 5-methylpyrazine-2-carboxylic acid 4-oxide, 5-methylpyrazinecarboxylic acid 4-oxide, 2-carboxy-5-methylpyrazine 4-oxide, 5-methyl-2-pyrazinecarboxylic acid 4-oxide, 5-methyl-4-oxy-pyrazine-2-carboxylic acid, 2-pyrazinecarboxylic acid, 5-methyl-, 4-oxide
PubChem CID 5310993
IUPAC Name 2-carboxy-6-methylpyrazin-1-ium-1-olate
SMILES CC1=CN=CC(C(O)=O)=[N+]1[O-]

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylpyrazine-2-carboxylic Acid 4-Oxide
RTECSNumber UQ2453000
TSCA No
Recommended Storage Refrigerator