accessibility menu, dialog, popup

UserName

CH3NNHOCl(E/Z)

CAS RN 51-15-0

IUPAC Name: (2E)-1-methyl-2-(nitrosomethylidene)-1,2-dihydropyridine hydrochloride
Synonyms: (2E)-1-methyl-2-(nitrosomethylidene)pyridine hydrochloride
Molecular Weight (g/mol): 172.61
Molecular Formula: C7H9ClN2O
InChi Key: HIGSLXSBYYMVKI-UHDJGPCESA-N
SMILES: Cl.CN1C=CC=C\C1=C/N=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)