accessibility menu, dialog, popup

UserName

CAS RN 50982-12-2

IUPAC Name: λ²-ruthenium(2+) (1Z,5Z)-cycloocta-1,5-diene dichloride
Synonyms: λ²-ruthenium(2+) 1,5-cyclooctadiene, (Z,Z)- dichloride
Molecular Weight (g/mol): 280.15
Molecular Formula: C8H12Cl2Ru
InChi Key: DMRVBCXRFYZCPR-PHFPKPIQSA-L
SMILES: [Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results