accessibility menu, dialog, popup

UserName

H3CNCH3OOONCH3CH3

CAS RN 509-34-2

IUPAC Name: 3',6'-bis(diethylamino)-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: solvent red 49
Molecular Weight (g/mol): 442.56
Molecular Formula: C28H30N2O3
InChi Key: DZNJMLVCIZGWSC-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C(OC3=CC(=CC=C3C22OC(=O)C3=CC=CC=C23)N(CC)CC)=C1
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)