CAS RN 50892-23-4
CAS RN 50892-23-4
IUPAC Name:
2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid
Synonyms:
pirinixic acid
Molecular Weight (g/mol):
323.8
Molecular Formula:
C14H14ClN3O2S
InChi Key:
SZRPDCCEHVWOJX-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C
1
–
1
of
1
results
1
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic Acid 99.0+%, TCI America™
CAS: 50892-23-4 Molecular Formula: C14H14ClN3O2S Molecular Weight (g/mol): 323.80 MDL Number: MFCD00191335 InChI Key: SZRPDCCEHVWOJX-UHFFFAOYSA-N Synonym: pirinixic acid,4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid,pirinixic acid inn,unii-86c4mrt55a,ccris 133,acide pirinixique inn-french,acido pirinixico inn-spanish,acidum pirinixicum inn-latin,acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio PubChem CID: 5694 ChEBI: CHEBI:32509 IUPAC Name: 2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid SMILES: CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C