missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic Acid 99.0+%, TCI America™
$266.56 - $803.41
Chemical Identifiers
| CAS | 50892-23-4 |
|---|---|
| Molecular Formula | C14H14ClN3O2S |
| Molecular Weight (g/mol) | 323.80 |
| MDL Number | MFCD00191335 |
| InChI Key | SZRPDCCEHVWOJX-UHFFFAOYSA-N |
| Synonym | pirinixic acid, 4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid, pirinixic acid inn, unii-86c4mrt55a, ccris 133, acide pirinixique inn-french, acido pirinixico inn-spanish, acidum pirinixicum inn-latin, acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio |
| PubChem CID | 5694 |
| ChEBI | CHEBI:32509 |
| IUPAC Name | 2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid |
| SMILES | CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C |
Chemical Identifiers
| 50892-23-4 | |
| 323.80 | |
| SZRPDCCEHVWOJX-UHFFFAOYSA-N | |
| 5694 | |
| 2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid |
| C14H14ClN3O2S | |
| MFCD00191335 | |
| pirinixic acid, 4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid, pirinixic acid inn, unii-86c4mrt55a, ccris 133, acide pirinixique inn-french, acido pirinixico inn-spanish, acidum pirinixicum inn-latin, acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio | |
| CHEBI:32509 | |
| CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C |
Specifications
| 155°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD00191335 | |
| SZRPDCCEHVWOJX-UHFFFAOYSA-N | |
| 2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid | |
| 5694 | |
| 323.80 | |
| [4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic Acid |
| 50892-23-4 | |
| 1 g | |
| C14H14ClN3O2S | |
| pirinixic acid, 4-chloro-6-2,3-xylidino-2-pyrimidinylthio acetic acid, pirinixic acid inn, unii-86c4mrt55a, ccris 133, acide pirinixique inn-french, acido pirinixico inn-spanish, acidum pirinixicum inn-latin, acetic acid, 4-chloro-6-2,3-dimethylphenyl amino-2-pyrimidinyl thio | |
| CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C | |
| 323.80 | |
| CHEBI:32509 | |
| ≥99.0% (HPLC,T) |
Safety and Handling
RTECSNumber : AG2915000
TSCA : No