accessibility menu, dialog, popup

UserName

ONON

CAS RN 5089-22-5

IUPAC Name: 2-[4-(1,3-benzoxazol-2-yl)naphthalen-1-yl]-1,3-benzoxazole
Synonyms: fluorescent brightener 367
Molecular Weight (g/mol): 362.39
Molecular Formula: C24H14N2O2
InChi Key: WFYSPVCBIJCZPX-UHFFFAOYSA-N
SMILES: O1C2=CC=CC=C2N=C1C1=C2C=CC=CC2=C(C=C1)C1=NC2=CC=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results