accessibility menu, dialog, popup

UserName

CH3NH2N

CAS RN 50670-64-9

IUPAC Name: 5-amino-2-methylbenzonitrile
Synonyms: 5-amino-2-methylbenzonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: YDZVQWCVKXYGIU-UHFFFAOYSA-N
SMILES: CC1=C(C=C(N)C=C1)C#N
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)