accessibility menu, dialog, popup

UserName

CAS RN 50650-59-4

IUPAC Name: 4-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 4-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 163.1
Molecular Formula: C6H4F3NO
InChi Key: IKHLLNMSMFVTLP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results