CAS RN 505083-04-5
IUPAC Name:
[3-fluoro-4-(methoxycarbonyl)phenyl]boronic acid
Synonyms:
3-fluoro-4-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol):
197.96
Molecular Formula:
C8H8BFO4
InChi Key:
YZYGXFXSMDUXJT-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C1F)B(O)O
1
–
2
of
2
results
1
Promotions Available
3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%
3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%
CAS: 505083-04-5 Molecular Formula: C8H8BFO4 Molecular Weight (g/mol): 197.956 MDL Number: MFCD04115665 InChI Key: YZYGXFXSMDUXJT-UHFFFAOYSA-N Synonym: 3-fluoro-4-methoxycarbonyl phenyl boronic acid,3-fluoro-4-methoxycarbonyl phenylboronic acid,3-fluoro-4-methoxycarbonyl benzeneboronic acid,methyl 4-borono-2-fluorobenzoate,4-methoxycarbonyl-3-fluorophenylboronic acid,pubchem17771,3-fluoro-4-methoxycarboxyphenylboronicacid,acmc-1atlt,3-fluoro-4-methoxycarbonyl-phenyl boronic acid PubChem CID: 2782818 IUPAC Name: (3-fluoro-4-methoxycarbonylphenyl)boronic acid SMILES: B(C1=CC(=C(C=C1)C(=O)OC)F)(O)O