missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10871G Supplier TCI America Supplier No. F10871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Fluoro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 505083-04-5
Melting Point 184°C
Color White
Molecular Formula C8H8BFO4
Quantity 1 g
MDL Number MFCD04115665
Synonym 3-fluoro-4-methoxycarbonyl phenyl boronic acid, 3-fluoro-4-methoxycarbonyl phenylboronic acid, 3-fluoro-4-methoxycarbonyl benzeneboronic acid, methyl 4-borono-2-fluorobenzoate, 4-methoxycarbonyl-3-fluorophenylboronic acid, pubchem17771, 3-fluoro-4-methoxycarboxyphenylboronicacid, acmc-1atlt, 3-fluoro-4-methoxycarbonyl-phenyl boronic acid
InChI Key YZYGXFXSMDUXJT-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C(=O)OC)F)(O)O
IUPAC Name (3-fluoro-4-methoxycarbonylphenyl)boronic acid
Molecular Weight (g/mol) 197.956
PubChem CID 2782818
Formula Weight 197.96
Physical Form Crystalline Powder
Show More Show Less