missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%

Catalog No. AAH5251006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5251006 5 g
AAH5251003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5251006 Supplier Thermo Scientific Chemicals Supplier No. H5251006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 505083-04-5 | C8H8BFO4 | 197.956 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 505083-04-5
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.956
MDL Number MFCD04115665
InChI Key YZYGXFXSMDUXJT-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxycarbonyl phenyl boronic acid, 3-fluoro-4-methoxycarbonyl phenylboronic acid, 3-fluoro-4-methoxycarbonyl benzeneboronic acid, methyl 4-borono-2-fluorobenzoate, 4-methoxycarbonyl-3-fluorophenylboronic acid, pubchem17771, 3-fluoro-4-methoxycarboxyphenylboronicacid, acmc-1atlt, 3-fluoro-4-methoxycarbonyl-phenyl boronic acid
PubChem CID 2782818
IUPAC Name (3-fluoro-4-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)OC)F)(O)O

Specifications

Melting Point 180°C to 184°C
Quantity 5 g
Formula Weight 197.96
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only