accessibility menu, dialog, popup

UserName

CAS RN 5027-32-7

CH3OH3COH3COCH3O

CAS RN 5027-32-7

IUPAC Name: 3,4-diacetylhexane-2,5-dione
Synonyms: 3,4-diacetylhexane-2,5-dione
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: CSKRBHOAJUMOKJ-UHFFFAOYSA-N
SMILES: CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)