accessibility menu, dialog, popup

UserName

CH3OHO

CAS RN 5020-41-7

IUPAC Name: 2-(3-methoxyphenyl)ethan-1-ol
Synonyms: 2-(3-methoxyphenyl)ethanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: UPPGEJSCUZMCMW-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CCO)=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)