accessibility menu, dialog, popup

UserName

H3CSCH3CH3

CAS RN 5008-73-1

IUPAC Name: 1-(propan-2-ylsulfanyl)propane
Synonyms: 1-(isopropylsulfanyl)propane
Molecular Weight (g/mol): 118.24
Molecular Formula: C6H14S
InChi Key: BDFDQOJJDDORSR-UHFFFAOYSA-N
SMILES: CCCSC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results