accessibility menu, dialog, popup

UserName

CAS RN 5006-62-2

IUPAC Name: ethyl piperidine-3-carboxylate
Synonyms: ethyl piperidine-3-carboxylate
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: XIWBSOUNZWSFKU-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCNC1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)