missing translation for 'onlineSavingsMsg'
Learn More

Ribose, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023004
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023004 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023004 Supplier Merck Emd Millipore Supplier No. PHR2101500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 50-69-1
Melting Point 88°C to 92°C (literature)
Molecular Formula C5H10O5
Linear Formula C5H10O5
Quantity 500 mg
MDL Number MFCD00135453
Synonym D-(-)-Ribose
Molecular Weight (g/mol) 150.13
Formula Weight 150.13
Grade Certified Reference Material
Physical Form Solid
Show More Show Less