CAS RN 4998-07-6
IUPAC Name:
4,5-dimethoxy-2-nitrobenzoate
Synonyms:
4,5-dimethoxy-2-nitrobenzoate
Molecular Weight (g/mol):
226.17
Molecular Formula:
C9H8NO6
InChi Key:
WWCMFGBGMJAJRX-UHFFFAOYSA-M
SMILES:
COC1=CC(C([O-])=O)=C(C=C1OC)[N+]([O-])=O
1
–
2
of
2
results
1
2
4,5-Dimethoxy-2-nitrobenzoic Acid 98.0+%, TCI America™
CAS: 4998-07-6 Molecular Formula: C9H8NO6 Molecular Weight (g/mol): 226.17 MDL Number: MFCD00014697 InChI Key: WWCMFGBGMJAJRX-UHFFFAOYSA-M Synonym: 4,5-dimethoxy-2-nitrobenzoic acid,6-nitroveratric acid,3,4-dimethoxy-6-nitrobenzoic acid,benzoic acid, 4,5-dimethoxy-2-nitro,zlchem 681,acmc-2097gn,ksc496i5r,4,5-dimethoxy-2-nitrobenzoicacid,2-nitro-4,5-dimethoxybenzoic acid,4,5-dimethoxy-2-nitro-benzoic acid PubChem CID: 78690 SMILES: COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC