missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic acid, 98%

Catalog No. AAL0851022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0851022 100 g
AAL0851006 5 g
AAL0851014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0851022 Supplier Thermo Scientific Chemicals Supplier No. L0851022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4998-07-6 | C9H8NO6 | 226.17 g/mol

4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.

Solubility
Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 4998-07-6
MDL Number MFCD00014697

Specifications

Melting Point 192°C to 195°C
Molecular Formula C9H8NO6
Quantity 100 g
Beilstein 2123903
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
Solubility Information Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
SMILES COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
Molecular Weight (g/mol) 226.17
PubChem CID 78690
Formula Weight 227.17
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-657-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only