missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. D26285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26285G 5 g
D262825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26285G Supplier TCI America Supplier No. D26285G

Chemical Identifiers

CAS 4998-07-6
Molecular Formula C9H8NO6
Molecular Weight (g/mol) 226.17
MDL Number MFCD00014697
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
PubChem CID 78690
SMILES COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC

Specifications

Melting Point 194°C
Quantity 5 g
Formula Weight 227.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzoic Acid