accessibility menu, dialog, popup

UserName

ClONCl

CAS RN 49609-84-9

IUPAC Name: 2-chloropyridine-3-carbonyl chloride
Synonyms: 2-chloropyridine-3-carbonyl chloride
Molecular Weight (g/mol): 176
Molecular Formula: C6H3Cl2NO
InChi Key: RXTRRIFWCJEMEL-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CN=C1Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)