accessibility menu, dialog, popup

UserName

H3COOClN

CAS RN 49608-01-7

IUPAC Name: ethyl 6-chloropyridine-3-carboxylate
Synonyms: ethyl 6-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: ILDJJTQWIZLGPO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Cl)N=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)